missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dimethoxybenzene, 98%

Catalog No. AAA1282222
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1282222 100 g
AAA1282236 500 g
AAA128220E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1282222 Supplier Thermo Scientific Chemicals Supplier No. A1282222
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 150-78-7 | C8H10O2 | 138.17 g/mol

1,4-Dimethoxybenzene is used as Pharmaceutical Intermediate. It is used in some paints and as a diazo dye. It is also used in perfumes and flavors in order to its floral odor. In addition, it is used on greasy skin, and with sulfur to treat acne, or as a dandruff treatment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Dimethoxybenzene is used as Pharmaceutical Intermediate. It is used in some paints and as a diazo dye. It is also used in perfumes and flavors in order to its floral odor. In addition, it is used on greasy skin, and with sulfur to treat acne, or as a dandruff treatment.

Solubility
Soluble in water(0.83g/L at 20°C) and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 150-78-7
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.17
MDL Number MFCD00008401
InChI Key OHBQPCCCRFSCAX-UHFFFAOYSA-N
Synonym p-dimethoxybenzene, p-methoxyanisole, hydroquinone dimethyl ether, benzene, 1,4-dimethoxy, quinol dimethyl ether, benzene, p-dimethoxy, dimethylhydroquinone ether, dimethyl ether hydroquinone, 1,4-dimethoxybenzol, usaf an-9
PubChem CID 9016
IUPAC Name 1,4-dimethoxybenzene
SMILES COC1=CC=C(OC)C=C1

Specifications

Melting Point 54°C to 58°C
Density 1.053
Boiling Point 212°C
Flash Point 94°C (201°F)
Odor Sweet
Quantity 100 g
Beilstein 774605
Sensitivity Light sensitive
Solubility Information Soluble in water(0.83g/L at 20°C) and methanol.
Formula Weight 138.17
Percent Purity 98%
Chemical Name or Material 1,4-Dimethoxybenzene
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315
Causes skin irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 205-771-9
RTECSNumber CZ6650000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only