missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dimethoxy-2-butyne 98.0+%, TCI America™

Catalog No. p-7128570
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D49511ML 1 mL
D49515ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D49511ML Supplier TCI America Supplier No. D49511ML

Chemical Identifiers

CAS 16356-02-8
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.14
MDL Number MFCD00048069
InChI Key YFUQYYGBJJCAPR-UHFFFAOYSA-N
PubChem CID 542354
IUPAC Name 1,4-dimethoxybut-2-yne
SMILES COCC#CCOC

Specifications

Melting Point -39°C
Color Yellow
Boiling Point 157°C
Quantity 1 mL
UN Number 3271
Formula Weight 114.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,4-Dimethoxy-2-butyne