accessibility menu, dialog, popup

UserName

CH3OOH3C

CAS RN 16356-02-8

IUPAC Name: 1,4-dimethoxybut-2-yne
Synonyms: 1,4-dimethoxybut-2-yne
Molecular Weight (g/mol): 114.14
Molecular Formula: C6H10O2
InChi Key: YFUQYYGBJJCAPR-UHFFFAOYSA-N
SMILES: COCC#CCOC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results