missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diisopropylbenzene 98.0+%, TCI America™

Catalog No. D092825ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D092825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D092825ML Supplier TCI America Supplier No. D092825ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 100-18-5
Molecular Formula C12H18
Molecular Weight (g/mol) 162.28
MDL Number MFCD00008892
InChI Key SPPWGCYEYAMHDT-UHFFFAOYSA-N
Synonym 1,4-diisopropylbenzene, p-diisopropylbenzene, benzene, p-diisopropyl, benzene, 1,4-bis 1-methylethyl, para-diisopropylbenzene, p-diisopropylbenzol, 1,4-bis 1-methylethyl benzene, unii-5m62031nzp, 1,4-di propan-2-yl benzene, dsstox_cid_6652
PubChem CID 7486
IUPAC Name 1,4-bis(propan-2-yl)benzene
SMILES CC(C)C1=CC=C(C=C1)C(C)C

Specifications

Color Colorless
Boiling Point 210°C
Quantity 25 mL
UN Number 3082
Formula Weight 162.28
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,4-Diisopropylbenzene
EINECSNumber (3)-0025
RTECSNumber CZ6360000
TSCA Yes