accessibility menu, dialog, popup

UserName

H3CCH3CH3H3C

CAS RN 100-18-5

IUPAC Name: 1,4-bis(propan-2-yl)benzene
Synonyms: diisopropylbenzene
Molecular Weight (g/mol): 162.28
Molecular Formula: C12H18
InChi Key: SPPWGCYEYAMHDT-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=C(C=C1)C(C)C
Boiling Point 
  • (3)
  • (4)
Percent Purity 
  • (1)
  • (6)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (6)
  • (3)
  • (4)
  • (1)
  • (6)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)