missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diisopropylbenzene, 99%

CAS: 100-18-5 | C12H18 | 162.28 g/mol

Supplier:  Thermo Scientific Chemicals 155210250

Quantity 25 g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC155210250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

100-18-5
162.28
SPPWGCYEYAMHDT-UHFFFAOYSA-N
7486
C12H18
MFCD00008892
1,4-diisopropylbenzene, p-diisopropylbenzene, benzene, p-diisopropyl, benzene, 1,4-bis 1-methylethyl, para-diisopropylbenzene, p-diisopropylbenzol, 1,4-bis 1-methylethyl benzene, unii-5m62031nzp, 1,4-di propan-2-yl benzene, dsstox_cid_6652
CC(C)C1=CC=C(C=C1)C(C)C

Specifications

1,4-Diisopropylbenzene
100-18-5
100.0
0.8600g/mL
76°C
Glass bottle
1.4890 to 1.4910
25 g
05,II,339
1,4-diisopropylbenzene, p-diisopropylbenzene, benzene, p-diisopropyl, benzene, 1,4-bis 1-methylethyl, para-diisopropylbenzene, p-diisopropylbenzol, 1,4-bis 1-methylethyl benzene, unii-5m62031nzp, 1,4-di propan-2-yl benzene, dsstox_cid_6652
SPPWGCYEYAMHDT-UHFFFAOYSA-N
1,4-di(propan-2-yl)benzene
7486
99%
99%
98.5
-17.0°C
203.0°C
98.5% min. (GC)
C12H18
C6H4[CH(CH3)2]2
MFCD00008892
0.86
Solubility in water: insoluble. Other solubilities: %iscible with alcohol, ether, benzene, acetone and, ccl4
CC(C)C1=CC=C(C=C1)C(C)C
162.28
162.27
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.

GHS P Statement
Wear protective gloves/protective clothing.
Wash face,hands and any exposed skin thoroughly after handling.

GHS Signal Word: Warning

EINECSNumber : 202-826-9

RTECSNumber : CZ6360000

TSCA : TSCA

SDS
Documents

Documents

Promotions

Promotions