missing translation for 'onlineSavingsMsg'
Learn More

1,4-Difluoro-2-iodobenzene 98.0+%, TCI America™

Catalog No. D35941G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35941G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D35941G Supplier TCI America Supplier No. D35941G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 2265-92-1
Boiling Point 183°C
Molecular Formula C6H3F2I
Quantity 1 g
MDL Number MFCD03094151
Synonym 2,5-difluoroiodobenzene, benzene,1,4-difluoro-2-iodo, 2,5-difluoro-1-iodobenzene, benzene, 1,4-difluoro-2-iodo, pubchem4402, acmc-209fxx, 1,4-bis fluoranyl-2-iodanyl-benzene
InChI Key WBYVNDUCUMNZPM-UHFFFAOYSA-N
SMILES FC1=CC=C(F)C(I)=C1
IUPAC Name 1,4-difluoro-2-iodobenzene
Molecular Weight (g/mol) 239.99
PubChem CID 223084
Formula Weight 239.99
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,4-Difluoro-2-iodobenzene
Show More Show Less