missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diethylpiperazine, 98%

CAS: 6483-50-7 | C8H18N2 | 142.246 g/mol

Supplier:  Thermo Scientific Chemicals B2474318

 View more versions of this product

Catalog No. AAB2474318


Only null left
Explore available promotions

Description

Description

1,4-Diethylpiperazine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Diethylpiperazine is used as pharmaceutical intermediate.

Solubility
Soluble in methanol, toluene.

Notes
store at a cool and dry place, with container tightly sealed. Store under nitrogen, recommended temperature is below 4°C.
Specifications

Specifications

1,4-Diethylpiperazine
C8H18N2
50 g
Soluble in methanol,toluene.
CCN1CCN(CC1)CC
142.246
142.24
6483-50-7
MFCD00126900
piperazine, 1,4-diethyl, 1,4-diethyl piperazine, piperazine,1,4-diethyl, acmc-1be2o
DDPRYTUJYNYJKV-UHFFFAOYSA-N
1,4-diethylpiperazine
80973
98%

Chemical Identifiers

6483-50-7
142.246
DDPRYTUJYNYJKV-UHFFFAOYSA-N
1,4-diethylpiperazine
C8H18N2
MFCD00126900
80973
CCN1CCN(CC1)CC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H226-H314-H335

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 229-341-5

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only