missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diethylpiperazine, 98%

Catalog No. p-7027747
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2474309 10 g
AAB2474318 50 g
AAB2474330 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2474309 Supplier Thermo Scientific Chemicals Supplier No. B2474309
Only null left

CAS: 6483-50-7 | C8H18N2 | 142.246 g/mol

1,4-Diethylpiperazine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Diethylpiperazine is used as pharmaceutical intermediate.

Solubility
Soluble in methanol, toluene.

Notes
store at a cool and dry place, with container tightly sealed. Store under nitrogen, recommended temperature is below 4°C.

Chemical Identifiers

CAS 6483-50-7
Molecular Formula C8H18N2
Molecular Weight (g/mol) 142.246
MDL Number MFCD00126900
InChI Key DDPRYTUJYNYJKV-UHFFFAOYSA-N
PubChem CID 80973
IUPAC Name 1,4-diethylpiperazine
SMILES CCN1CCN(CC1)CC

Specifications

Quantity 10 g
Synonym piperazine, 1,4-diethyl, 1,4-diethyl piperazine, piperazine,1,4-diethyl, acmc-1be2o
Solubility Information Soluble in methanol,toluene.
Formula Weight 142.24
Percent Purity 98%
Chemical Name or Material 1,4-Diethylpiperazine
Hazard Category H226-H314-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 229-341-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only