missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diethylbenzene 98.0+%, TCI America™

Catalog No. p-7126238
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D04795ML 5 mL
D047925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D04795ML Supplier TCI America Supplier No. D04795ML

Chemical Identifiers

CAS 105-05-5
Molecular Formula C10H14
Molecular Weight (g/mol) 134.22
MDL Number MFCD00009264
InChI Key DSNHSQKRULAAEI-UHFFFAOYSA-N
Synonym p-diethylbenzene, benzene, 1,4-diethyl, p-ethylethylbenzene, benzene, p-diethyl, para-diethylbenzene, unii-0psm16x42d, acmc-1bu3z, dsstox_cid_6711, dsstox_rid_78192, dsstox_gsid_26711
PubChem CID 7734
ChEBI CHEBI:34062
IUPAC Name 1,4-diethylbenzene
SMILES CCC1=CC=C(CC)C=C1

Specifications

Melting Point -43°C
Color Colorless
Boiling Point 184°C
Quantity 5 mL
UN Number 2049
Formula Weight 134.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,4-Diethylbenzene
EINECSNumber (3)-0013&(3)-0060
RTECSNumber CZ5640700
TSCA Yes