accessibility menu, dialog, popup

UserName

CAS RN 105-05-5

IUPAC Name: 1,4-diethylbenzene
Synonyms: diethylbenzene P-diethylbenzene
Molecular Weight (g/mol): 134.22
Molecular Formula: C10H14
InChi Key: DSNHSQKRULAAEI-UHFFFAOYSA-N
SMILES: CCC1=CC=C(CC)C=C1
Boiling Point 
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (3)
  • (4)
  • (2)
  • (5)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (4)