missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diethylbenzene, 98%

Catalog No. AAA1334306
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1334306 5 g
AAA1334314 25 g
AAA1334322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1334306 Supplier Thermo Scientific Chemicals Supplier No. A1334306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 105-05-5 | C10H14 | 134.22 g/mol

1,4-Diethylbenzene, is used as a solvent and an intermediate in the manufacture of other organic compounds for the finished products of such as curing agents, engineering plastics andcross-linking agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Diethylbenzene, is used as a solvent and an intermediate in the manufacture of other organic compounds for the finished products of such as curing agents, engineering plastics andcross-linking agents.

Solubility
It is insoluble in water. But soluble in ethanol, benzene,carbon tetrachloride.

Notes
Stable under ordinary conditions. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 105-05-5
Molecular Formula C10H14
Molecular Weight (g/mol) 134.22
MDL Number MFCD00009264
InChI Key DSNHSQKRULAAEI-UHFFFAOYSA-N
Synonym p-diethylbenzene, benzene, 1,4-diethyl, p-ethylethylbenzene, benzene, p-diethyl, para-diethylbenzene, unii-0psm16x42d, acmc-1bu3z, dsstox_cid_6711, dsstox_rid_78192, dsstox_gsid_26711
PubChem CID 7734
ChEBI CHEBI:34062
IUPAC Name 1,4-diethylbenzene
SMILES CCC1=CC=C(CC)C=C1

Specifications

Melting Point -43°C
Density 0.862
Boiling Point 182°C to 184°C
Flash Point 55°C (131°F)
Odor Aromatic
Refractive Index 1.495
Quantity 5 g
UN Number UN2049
Beilstein 1903396
Solubility Information It is insoluble in water. But soluble in ethanol,benzene,carbon tetrachloride.
Formula Weight 134.22
Percent Purity 98%
Chemical Name or Material 1,4-Diethylbenzene
Show More Show Less
Hazard Category H226-H304-H315-H318
Hazard Statement GHS H Statement
H226
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P280-P301+P310-P302+P352-P305+P351+P338-P310-P331-P332+P313-P362-P374-P380-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: DIETHYLBENZENE
EINECSNumber 203-265-2
RTECSNumber CZ5640700
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only