missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dicyclohexylbenzene 98.0+%, TCI America™

Catalog No. D102825G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D102825G 25 g
D1028250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D102825G Supplier TCI America Supplier No. D102825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1087-02-1
Molecular Formula C18H26
Molecular Weight (g/mol) 242.406
MDL Number MFCD00001452
InChI Key QQFSIGWYINAJOB-UHFFFAOYSA-N
Synonym p-dicyclohexylbenzene, unii-a4m5u01mcy, a4m5u01mcy, benzene, 1,4-dicyclohexyl, p-dicyclohexylbenzol, benzene,4-dicyclohexyl, benzene, p-dicyclohexyl, p-dicyclohexyl benzene, qqfsigwyinajob-uhfffaoysa, unii-n5v9zbl13v component
PubChem CID 70664
IUPAC Name 1,4-dicyclohexylbenzene
SMILES C1CCC(CC1)C2=CC=C(C=C2)C3CCCCC3

Specifications

Melting Point 102°C
Color White
Boiling Point 195°C
Quantity 25 g
Formula Weight 242.41
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Dicyclohexylbenzene
EINECSNumber (4)-0041
TSCA No