missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dichloroanthraquinone 98.0+%, TCI America™

Catalog No. p-7127346
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D25181G 1 g
D25185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D25181G Supplier TCI America Supplier No. D25181G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 602-25-5
Molecular Formula C14H6Cl2O2
Molecular Weight (g/mol) 277.10
MDL Number MFCD00192002
InChI Key CAHGWVAXFJXDNI-UHFFFAOYSA-N
PubChem CID 4179424
IUPAC Name 1,4-dichloro-9,10-dihydroanthracene-9,10-dione
SMILES ClC1=C2C(=O)C3=CC=CC=C3C(=O)C2=C(Cl)C=C1

Specifications

Melting Point 190°C
Quantity 1 g
Formula Weight 277.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Dichloroanthraquinone