missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dichloro-2-nitrobenzene 99.0+%, TCI America™

Catalog No. D038725G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D038725G 25 g
D0387500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D038725G Supplier TCI America Supplier No. D038725G

Chemical Identifiers

CAS 89-61-2
Molecular Formula C6H3Cl2NO2
Molecular Weight (g/mol) 192.00
MDL Number MFCD00007074
InChI Key RZKKOBGFCAHLCZ-UHFFFAOYSA-N
Synonym 2,5-dichloronitrobenzene, nitro-p-dichlorobenzene, benzene, 1,4-dichloro-2-nitro, 1-nitro-2,5-dichlorobenzene, 2,5-dichlornitrobenzen, 2,5-dichloro-1-nitrobenzene, benzene, 2,5-dichloronitro, 2,5-dichloronitrobenzol, unii-iy18g50ff4, ccris 3096
PubChem CID 6977
IUPAC Name 1,4-dichloro-2-nitrobenzene
SMILES [O-][N+](=O)C1=CC(Cl)=CC=C1Cl

Specifications

Melting Point 55°C
Boiling Point 98°C
Quantity 25 g
UN Number 3077
Formula Weight 192.00
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Dichloro-2-nitrobenzene
EINECSNumber (3)-0455
RTECSNumber CZ5260000
TSCA Yes