missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Dichloro-2-butyne 97.0+%, TCI America™
$61.61 - $192.52
Chemical Identifiers
| CAS | 821-10-3 |
|---|---|
| Molecular Formula | C4H4Cl2 |
| Molecular Weight (g/mol) | 122.98 |
| MDL Number | MFCD00000981 |
| InChI Key | RCHDLEVSZBOHOS-UHFFFAOYSA-N |
| Synonym | 1,4-dichloro-2-butyne, 2-butyne, 1,4-dichloro, 1,4-dichlorobutyne, 2-butyne,4-dichloro, 1,4 dichloro-2-butyne, 1,4-dichloro-2-butine, 1,4-dichloro-but-2-yne, acmc-209pn5, wln: g2uu2g, 4-01-00-00973 beilstein handbook reference |
| PubChem CID | 13182 |
| IUPAC Name | 1,4-dichlorobut-2-yne |
| SMILES | ClCC#CCCl |
Chemical Identifiers
| 821-10-3 | |
| 122.98 | |
| RCHDLEVSZBOHOS-UHFFFAOYSA-N | |
| 13182 | |
| ClCC#CCCl |
| C4H4Cl2 | |
| MFCD00000981 | |
| 1,4-dichloro-2-butyne, 2-butyne, 1,4-dichloro, 1,4-dichlorobutyne, 2-butyne,4-dichloro, 1,4 dichloro-2-butyne, 1,4-dichloro-2-butine, 1,4-dichloro-but-2-yne, acmc-209pn5, wln: g2uu2g, 4-01-00-00973 beilstein handbook reference | |
| 1,4-dichlorobut-2-yne |
Specifications
| 821-10-3 | |
| 166°C | |
| MFCD00000981 | |
| 2810 | |
| RCHDLEVSZBOHOS-UHFFFAOYSA-N | |
| 1,4-dichlorobut-2-yne | |
| 13182 | |
| ≥97.0% (GC) | |
| 1,4-Dichloro-2-butyne |
| Yellow | |
| C4H4Cl2 | |
| 5 g | |
| 1,4-dichloro-2-butyne, 2-butyne, 1,4-dichloro, 1,4-dichlorobutyne, 2-butyne,4-dichloro, 1,4 dichloro-2-butyne, 1,4-dichloro-2-butine, 1,4-dichloro-but-2-yne, acmc-209pn5, wln: g2uu2g, 4-01-00-00973 beilstein handbook reference | |
| ClCC#CCCl | |
| 122.98 | |
| 122.98 | |
| Liquid |
Safety and Handling
RTECSNumber : ER9600000
TSCA : Yes
Recommended Storage : Refrigerator