missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dichloro-2-butyne 97.0+%, TCI America™

Catalog No. d15285g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
D15285G 5 g
D152825G 25 g
2 options

Catalog No. D15285G

Supplier: TCI America D15285G

Only null left

Chemical Identifiers

CAS 821-10-3
Molecular Formula C4H4Cl2
Molecular Weight (g/mol) 122.98
MDL Number MFCD00000981
InChI Key RCHDLEVSZBOHOS-UHFFFAOYSA-N
Synonym 1,4-dichloro-2-butyne, 2-butyne, 1,4-dichloro, 1,4-dichlorobutyne, 2-butyne,4-dichloro, 1,4 dichloro-2-butyne, 1,4-dichloro-2-butine, 1,4-dichloro-but-2-yne, acmc-209pn5, wln: g2uu2g, 4-01-00-00973 beilstein handbook reference
PubChem CID 13182
IUPAC Name 1,4-dichlorobut-2-yne
SMILES ClCC#CCCl

Specifications

Color Yellow
Boiling Point 166°C
Quantity 5 g
UN Number 2810
Formula Weight 122.98
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,4-Dichloro-2-butyne
RTECSNumber ER9600000
TSCA Yes
Recommended Storage Refrigerator