missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dibutylbenzene 98.0+%, TCI America™

Catalog No. D27815ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D27815ML 5 mL
D278125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D27815ML Supplier TCI America Supplier No. D27815ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1571-86-4
Molecular Formula C14H22
Molecular Weight (g/mol) 190.33
MDL Number MFCD01076591
InChI Key VGQOZYOOFXEGDA-UHFFFAOYSA-N
Synonym 1,4-di-n-butylbenzene, benzene, 1,4-dibutyl, p-dibutylbenzol, p-dibutylbenzene, acmc-209dfm, p-di-tert-butyl benzene
PubChem CID 519170
IUPAC Name 1,4-dibutylbenzene
SMILES CCCCC1=CC=C(CCCC)C=C1

Specifications

Color Colorless
Quantity 5 mL
Formula Weight 190.33
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,4-Dibutylbenzene
EINECSNumber (3)-0025
TSCA No