accessibility menu, dialog, popup

UserName

H3CCH3

CAS RN 1571-86-4

IUPAC Name: 1,4-dibutylbenzene
Synonyms: 1,4-dibutylbenzene
Molecular Weight (g/mol): 190.33
Molecular Formula: C14H22
InChi Key: VGQOZYOOFXEGDA-UHFFFAOYSA-N
SMILES: CCCCC1=CC=C(CCCC)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)