missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Dibromonaphthalene 98.0+%, TCI America™
Supplier: TCI America D23595G
Specifications
| 1,4-Dibromonaphthalene | |
| 82°C | |
| C10H6Br2 | |
| 5 g | |
| IBGUDZMIAZLJNY-UHFFFAOYSA-N | |
| 1,4-dibromonaphthalene | |
| 66521 | |
| ≥98.0% (GC) |
| 83-53-4 | |
| White-Yellow | |
| MFCD00041823 | |
| naphthalene, 1,4-dibromo, 1,4-dibromonapthalene, 1.4-dibromonaphthalene, acmc-209psk, 1,4-dibromo-naphthalen, l ,4-dibromonaphthalene, ksc490o3f, 1,4-dibromonaphthalene 5g | |
| BrC1=C2C=CC=CC2=C(Br)C=C1 | |
| 285.97 | |
| 285.97 | |
| Crystalline Powder |
Chemical Identifiers
| 83-53-4 | |
| 285.97 | |
| IBGUDZMIAZLJNY-UHFFFAOYSA-N | |
| 66521 | |
| BrC1=C2C=CC=CC2=C(Br)C=C1 |
| C10H6Br2 | |
| MFCD00041823 | |
| naphthalene, 1,4-dibromo, 1,4-dibromonapthalene, 1.4-dibromonaphthalene, acmc-209psk, 1,4-dibromo-naphthalen, l ,4-dibromonaphthalene, ksc490o3f, 1,4-dibromonaphthalene 5g | |
| 1,4-dibromonaphthalene |
Safety and Handling
TSCA : Yes