accessibility menu, dialog, popup

UserName

CAS RN 83-53-4

IUPAC Name: 1,4-dibromonaphthalene
Synonyms: naphthalene, 1,4-dibromo-
Molecular Weight (g/mol): 285.97
Molecular Formula: C10H6Br2
InChi Key: IBGUDZMIAZLJNY-UHFFFAOYSA-N
SMILES: BrC1=C2C=CC=CC2=C(Br)C=C1
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)