missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Dibromonaphthalene, 98+%
CAS: 83-53-4 | C10H6Br2 | 285.97 g/mol
$610.14 - $610.14
Chemical Identifiers
| CAS | 83-53-4 |
|---|---|
| Molecular Formula | C10H6Br2 |
| Molecular Weight (g/mol) | 285.97 |
| MDL Number | MFCD00041823 |
| InChI Key | IBGUDZMIAZLJNY-UHFFFAOYSA-N |
| Synonym | naphthalene, 1,4-dibromo, 1,4-dibromonapthalene, 1.4-dibromonaphthalene, acmc-209psk, 1,4-dibromo-naphthalen, l ,4-dibromonaphthalene, ksc490o3f, 1,4-dibromonaphthalene 5g |
| PubChem CID | 66521 |
| IUPAC Name | 1,4-dibromonaphthalene |
| SMILES | BrC1=C2C=CC=CC2=C(Br)C=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC406500250
|
Thermo Scientific Chemicals
406500250 |
25 g | Glass bottle |
Each for $610.14
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 83-53-4 | |
| 285.97 | |
| IBGUDZMIAZLJNY-UHFFFAOYSA-N | |
| 66521 | |
| BrC1=C2C=CC=CC2=C(Br)C=C1 |
| C10H6Br2 | |
| MFCD00041823 | |
| naphthalene, 1,4-dibromo, 1,4-dibromonapthalene, 1.4-dibromonaphthalene, acmc-209psk, 1,4-dibromo-naphthalen, l ,4-dibromonaphthalene, ksc490o3f, 1,4-dibromonaphthalene 5g | |
| 1,4-dibromonaphthalene |
Specifications
| 83-53-4 | |
| 100.0 | |
| White to Beige | |
| 98% min. (GC) | |
| C10H6Br2 | |
| 25 g | |
| Solubility in water: insoluble | |
| BrC1=C2C=CC=CC2=C(Br)C=C1 | |
| 285.97 | |
| 285.96 | |
| Crystalline Powder or Crystals |
| 98.0 | |
| 80.0°C to 82.0°C | |
| Authentic | |
| Glass bottle | |
| MFCD00041823 | |
| naphthalene, 1,4-dibromo, 1,4-dibromonapthalene, 1.4-dibromonaphthalene, acmc-209psk, 1,4-dibromo-naphthalen, l ,4-dibromonaphthalene, ksc490o3f, 1,4-dibromonaphthalene 5g | |
| IBGUDZMIAZLJNY-UHFFFAOYSA-N | |
| 1,4-dibromonaphthalene | |
| 66521 | |
| 98+% | |
| 1, 4-Dibromonaphthalene, 99% |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 201-484-8
TSCA : TSCA
RUO – Research Use Only