missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dibromonaphthalene, 98%

Catalog No. AAA1817106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1817106 5 g
AAA1817114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1817106 Supplier Thermo Scientific Chemicals Supplier No. A1817106

CAS: 83-53-4 | C10H6Br2 | 285.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 83-53-4
Molecular Formula C10H6Br2
Molecular Weight (g/mol) 285.97
MDL Number MFCD00041823
InChI Key IBGUDZMIAZLJNY-UHFFFAOYSA-N
Synonym naphthalene, 1,4-dibromo, 1,4-dibromonapthalene, 1.4-dibromonaphthalene, acmc-209psk, 1,4-dibromo-naphthalen, l ,4-dibromonaphthalene, ksc490o3f, 1,4-dibromonaphthalene 5g
PubChem CID 66521
IUPAC Name 1,4-dibromonaphthalene
SMILES BrC1=C2C=CC=CC2=C(Br)C=C1

Specifications

Melting Point 80°C to 82°C
Quantity 5 g
Formula Weight 285.97
Percent Purity 98%
Chemical Name or Material 1,4-Dibromonaphthalene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 201-484-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only