missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dibromobutane, 99%

Catalog No. AC161281000
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC161281000 100 mL Glass bottle
AC161285000 500 mL Glass bottle
AC161280010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC161281000 Supplier Thermo Scientific Chemicals Supplier No. 161281000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 110-52-1 | C4H8Br2 | 215.92 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 110-52-1
Molecular Formula C4H8Br2
Molecular Weight (g/mol) 215.92
MDL Number MFCD00000261
InChI Key ULTHEAFYOOPTTB-UHFFFAOYSA-N
Synonym tetramethylene dibromide, butane, 1,4-dibromo, 1,4-butylene bromide, tetramethylene bromide, 1,4-dibrombutan, tetramethylenebromide, alpha,omega-dibromobutane, unii-g49phr6jfu, 1,4-dibrombutan german, 1,4-dibromo-butane
PubChem CID 8056
IUPAC Name 1,4-dibromobutane
SMILES C(CCBr)CBr

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -20°C
Color Brown-Yellow to Colorless
Density 1.8080g/mL
Boiling Point 197°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5176 to 1.5196
Linear Formula Br(CH2)4Br
Quantity 100 mL
Beilstein 01, 120
Fieser 13,313
Specific Gravity 1.808
Solubility Information Solubility in water: immiscible. Other solubilities: soluble in all usual organic solvents and alcohols,soluble in chloroform
Formula Weight 215.92
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1, 4-Dibromobutane, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Toxic if swallowed.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
EINECSNumber 203-775-5
RTECSNumber EJ7565000
TSCA TSCA

RUO – Research Use Only