missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dibromo-2-iodobenzene 98.0+%, TCI America™

Catalog No. D49065G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D49065G 5 g
D49061G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D49065G Supplier TCI America Supplier No. D49065G
Only null left

Chemical Identifiers

CAS 89284-52-6
Molecular Formula C6H3Br2I
Molecular Weight (g/mol) 361.802
MDL Number MFCD07778993
InChI Key VLRYPRKTXDPVNN-UHFFFAOYSA-N
PubChem CID 13406348
IUPAC Name 1,4-dibromo-2-iodobenzene
SMILES C1=CC(=C(C=C1Br)I)Br

Specifications

Melting Point 41°C
Boiling Point 180°C
Quantity 5 g
Formula Weight 361.80
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Dibromo-2-iodobenzene