missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dibromo-2-fluorobenzene 98.0+%, TCI America™

Catalog No. D221225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D221225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D221225G Supplier TCI America Supplier No. D221225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1435-52-5
Molecular Formula C6H3Br2F
Molecular Weight (g/mol) 253.896
MDL Number MFCD00010603
InChI Key WNSNPGHNIJOOPM-UHFFFAOYSA-N
Synonym 2,5-dibromo-fluorobenzene, 2-fluoro-4-bromobromobenzene, 4-bromo-2-fluorobromobenzene, 2,5-dibromofluorobenzene, benzene, 1,4-dibromo-2-fluoro, 1,4-dibromo-2-fluorobenzene, 2,5-dibromo-1-fluorobenzene, pubchem3448, ksc494g9h, 1,4-dibromo-2-fluoro-benzene
PubChem CID 137004
IUPAC Name 1,4-dibromo-2-fluorobenzene
SMILES C1=CC(=C(C=C1Br)F)Br

Specifications

Quantity 25 g
Formula Weight 253.90
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material 1,4-Dibromo-2-fluorobenzene