missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dibromo-2-fluorobenzene, 98%

Catalog No. AAB2128114
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2128114 25 g
AAB2128122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2128114 Supplier Thermo Scientific Chemicals Supplier No. B2128114

CAS: 1435-52-5 | C6H3Br2F | 253.896 g/mol

It is used in preparation of 1,4-bis(2-hydroxy-2-methyl-3-butynyl)-2-fluorobenzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in preparation of 1,4-bis(2-hydroxy-2-methyl-3-butynyl)-2-fluorobenzene.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 1435-52-5
Molecular Formula C6H3Br2F
Molecular Weight (g/mol) 253.896
MDL Number MFCD00010603
InChI Key WNSNPGHNIJOOPM-UHFFFAOYSA-N
Synonym 2,5-dibromo-fluorobenzene, 2-fluoro-4-bromobromobenzene, 4-bromo-2-fluorobromobenzene, 2,5-dibromofluorobenzene, benzene, 1,4-dibromo-2-fluoro, 1,4-dibromo-2-fluorobenzene, 2,5-dibromo-1-fluorobenzene, pubchem3448, ksc494g9h, 1,4-dibromo-2-fluoro-benzene
PubChem CID 137004
IUPAC Name 1,4-dibromo-2-fluorobenzene
SMILES C1=CC(=C(C=C1Br)F)Br

Specifications

Melting Point 33°C to 35°C
Boiling Point 216°C
Flash Point 101°C (213°F)
Quantity 25 g
Beilstein 3236451
Solubility Information Insoluble in water.
Formula Weight 253.91
Percent Purity 98%
Chemical Name or Material 1,4-Dibromo-2-fluorobenzene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only