missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dibromo-2-butanol 95.0+%, TCI America™

Catalog No. D17435G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D17435G 5 g
D174325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D17435G Supplier TCI America Supplier No. D17435G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 19398-47-1
Molecular Formula C4H8Br2O
Molecular Weight (g/mol) 231.915
MDL Number MFCD00000252
InChI Key PSSRAPMBSMSACN-UHFFFAOYSA-N
Synonym 1,4-dibromo-2-butanol, 2-butanol, 1,4-dibromo, 2-butanol,1,4-dibromo, 2-butanol,4-dibromo, acmc-1b9jv, 1,4-dibromo-butan-2-ol, wln: e2yq1e, 4-01-00-01581 beilstein handbook reference, racemic, 1 4-dibromo-2-hydroxybutane
PubChem CID 29570
IUPAC Name 1,4-dibromobutan-2-ol
SMILES C(CBr)C(CBr)O

Specifications

Boiling Point 118°C
Quantity 5 g
Formula Weight 231.92
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,4-Dibromo-2-butanol
RTECSNumber EL1515000
TSCA No