CAS RN 19398-47-1
IUPAC Name:
1,4-dibromobutan-2-ol
Synonyms:
2-butanol, 1,4-dibromo-
Molecular Weight (g/mol):
231.92
Molecular Formula:
C4H8Br2O
InChi Key:
PSSRAPMBSMSACN-UHFFFAOYNA-N
SMILES:
OC(CBr)CCBr
1
–
1
of
1
results
1
1,4-Dibromo-2-butanol 95.0+%, TCI America™
CAS: 19398-47-1 Molecular Formula: C4H8Br2O Molecular Weight (g/mol): 231.915 MDL Number: MFCD00000252 InChI Key: PSSRAPMBSMSACN-UHFFFAOYSA-N Synonym: 1,4-dibromo-2-butanol,2-butanol, 1,4-dibromo,2-butanol,1,4-dibromo,2-butanol,4-dibromo,acmc-1b9jv,1,4-dibromo-butan-2-ol,wln: e2yq1e,4-01-00-01581 beilstein handbook reference,racemic,1 4-dibromo-2-hydroxybutane PubChem CID: 29570 IUPAC Name: 1,4-dibromobutan-2-ol SMILES: C(CBr)C(CBr)O