missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dibromo-2,5-dimethoxybenzene, 98+%

CAS: 2674-34-2 | C8H8Br2O2 | 295.958 g/mol

Supplier:  Thermo Scientific Chemicals A1985809

 View more versions of this product

Catalog No. AAA1985809


Only null left
Explore available promotions

Description

Description

It is employed as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

1,4-Dibromo-2,5-dimethoxybenzene
144°C to 148°C
MFCD00025694
1953103
Insoluble in water.
COC1=CC(=C(C=C1Br)OC)Br
295.958
295.96
2674-34-2
C8H8Br2O2
10 g
2,5-dibromo-1,4-dimethoxybenzene, 1,4-dibromo-2,5-dimethoxy-benzene, 2,5-dibromohydroquinone dimethyl ether, benzene, 1,4-dibromo-2,5-dimethoxy, pubchem16840, acmc-209gsv, maybridge1_005131, ksc496i0h, 2,5-dibromo-4-methoxyanisole, 1,4-dibromo-3,6-dimethoxybenzene
CHCLRVOURKGRSW-UHFFFAOYSA-N
1,4-dibromo-2,5-dimethoxybenzene
231240
≥98%

Chemical Identifiers

2674-34-2
295.958
CHCLRVOURKGRSW-UHFFFAOYSA-N
231240
COC1=CC(=C(C=C1Br)OC)Br
C8H8Br2O2
MFCD00025694
2,5-dibromo-1,4-dimethoxybenzene, 1,4-dibromo-2,5-dimethoxy-benzene, 2,5-dibromohydroquinone dimethyl ether, benzene, 1,4-dibromo-2,5-dimethoxy, pubchem16840, acmc-209gsv, maybridge1_005131, ksc496i0h, 2,5-dibromo-4-methoxyanisole, 1,4-dibromo-3,6-dimethoxybenzene
1,4-dibromo-2,5-dimethoxybenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only