missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Dibromo-2,5-bis(bromomethyl)benzene 97.0+%, TCI America™
$171.49 - $557.35
Chemical Identifiers
| CAS | 35335-16-1 |
|---|---|
| Molecular Formula | C8H6Br4 |
| Molecular Weight (g/mol) | 421.75 |
| MDL Number | MFCD03411745 |
| InChI Key | UOCMTKLYRROETA-UHFFFAOYSA-N |
| Synonym | alpha,alpha,2,5-Tetrabromo-p-xylene |
| PubChem CID | 640222 |
| IUPAC Name | 1,4-dibromo-2,5-bis(bromomethyl)benzene |
| SMILES | BrCC1=CC(Br)=C(CBr)C=C1Br |
Chemical Identifiers
| 35335-16-1 | |
| 421.75 | |
| UOCMTKLYRROETA-UHFFFAOYSA-N | |
| 640222 | |
| BrCC1=CC(Br)=C(CBr)C=C1Br |
| C8H6Br4 | |
| MFCD03411745 | |
| alpha,alpha,2,5-Tetrabromo-p-xylene | |
| 1,4-dibromo-2,5-bis(bromomethyl)benzene |
Specifications
| 35335-16-1 | |
| White-Yellow | |
| MFCD03411745 | |
| 3261 | |
| UOCMTKLYRROETA-UHFFFAOYSA-N | |
| 1,4-dibromo-2,5-bis(bromomethyl)benzene | |
| 640222 | |
| ≥97.0% (GC) | |
| 1,4-Dibromo-2,5-bis(bromomethyl)benzene |
| 160°C | |
| C8H6Br4 | |
| 1 g | |
| alpha,alpha,2,5-Tetrabromo-p-xylene | |
| BrCC1=CC(Br)=C(CBr)C=C1Br | |
| 421.75 | |
| 421.75 | |
| Crystalline Powder |
Safety and Handling
TSCA : No