missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Dibromo-2,3,5,6-tetramethylbenzene 98.0+%, TCI America™
$141.67 - $413.83
Chemical Identifiers
| CAS | 1646-54-4 |
|---|---|
| Molecular Formula | C10H12Br2 |
| Molecular Weight (g/mol) | 292.014 |
| MDL Number | MFCD00017787 |
| InChI Key | IEWAQZBDJDWANM-UHFFFAOYSA-N |
| Synonym | 3,6-Dibromodurene |
| PubChem CID | 74241 |
| IUPAC Name | 1,4-dibromo-2,3,5,6-tetramethylbenzene |
| SMILES | CC1=C(C(=C(C(=C1Br)C)C)Br)C |
Chemical Identifiers
| 1646-54-4 | |
| 292.014 | |
| IEWAQZBDJDWANM-UHFFFAOYSA-N | |
| 74241 | |
| CC1=C(C(=C(C(=C1Br)C)C)Br)C |
| C10H12Br2 | |
| MFCD00017787 | |
| 3,6-Dibromodurene | |
| 1,4-dibromo-2,3,5,6-tetramethylbenzene |
Specifications
| 1646-54-4 | |
| White | |
| C10H12Br2 | |
| 1 g | |
| IEWAQZBDJDWANM-UHFFFAOYSA-N | |
| 1,4-dibromo-2,3,5,6-tetramethylbenzene | |
| 74241 | |
| ≥98.0% (GC) | |
| 1,4-Dibromo-2,3,5,6-tetramethylbenzene |
| 203°C | |
| 317°C | |
| MFCD00017787 | |
| 3,6-Dibromodurene | |
| CC1=C(C(=C(C(=C1Br)C)C)Br)C | |
| 292.014 | |
| 292.01 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes