accessibility menu, dialog, popup

UserName

CAS RN 1646-54-4

CH3CH3BrCH3H3CBr

CAS RN 1646-54-4

IUPAC Name: 1,4-dibromo-2,3,5,6-tetramethylbenzene
Synonyms: 1,4-dibromo-2,3,5,6-tetramethylbenzene
Molecular Weight (g/mol): 292.01
Molecular Formula: C10H12Br2
InChi Key: IEWAQZBDJDWANM-UHFFFAOYSA-N
SMILES: CC1=C(C)C(Br)=C(C)C(C)=C1Br
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results