missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Dibromo-2,3,5,6-tetramethylbenzene 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 203°C |
| Color | White |
| Boiling Point | 317°C |
| Quantity | 5 g |
| Formula Weight | 292.01 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1,4-Dibromo-2,3,5,6-tetramethylbenzene |
Safety and Handling
| TSCA | Yes |