missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dibromo-2,3,5,6-tetramethylbenzene 98.0+%, TCI America™

Catalog No. D51681G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D51681G 1 g
D51685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D51681G Supplier TCI America Supplier No. D51681G
Only null left

Chemical Identifiers

CAS 1646-54-4
Molecular Formula C10H12Br2
Molecular Weight (g/mol) 292.014
MDL Number MFCD00017787
InChI Key IEWAQZBDJDWANM-UHFFFAOYSA-N
Synonym 3,6-Dibromodurene
PubChem CID 74241
IUPAC Name 1,4-dibromo-2,3,5,6-tetramethylbenzene
SMILES CC1=C(C(=C(C(=C1Br)C)C)Br)C

Specifications

Melting Point 203°C
Color White
Boiling Point 317°C
Quantity 1 g
Formula Weight 292.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Dibromo-2,3,5,6-tetramethylbenzene