Learn More
(+/-)-1,4-Diazabicyclo[4.4.0]decane, 98+%
CAS: 4430-75-5 | C8H16N2 | 140.23 g/mol
Supplier: Thermo Scientific Chemicals H5230503
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| (+/-)-1,4-Diazabicyclo[4.4.0]decane | |
| 68°C to 72°C (3 mmHg) | |
| MFCD01098257 | |
| UN3267 | |
| ONHPOXROAPYCGT-UHFFFAOYNA-N | |
| octahydro-1H-pyrido[1,2-a]pyrazine | |
| 2876391 | |
| ≥98% |
| 4430-75-5 | |
| C8H16N2 | |
| 1 g | |
| octahydro-1h-pyrido 1,2-a pyrazine, octahydro-2h-pyrido 1,2-a pyrazine, octahydro-pyrido 1,2-a pyrazine, 1,4-diazabicyclo 4.4.0 decane, octahydro-1h-pyrido 1,2-a piperazine, chembl98511, octahydropyrido 1,2-a pyrazine, 2,3,4,6,7,8,9,9a-octahydro-1h-pyrido 1,2-a pyrazine, +/--1,4-diazabicyclo, octahydropyrido 1,2-a-pyrazine | |
| C1CCN2CCNCC2C1 | |
| 140.23 | |
| 140.23 |
Chemical Identifiers
| 4430-75-5 | |
| 140.23 | |
| ONHPOXROAPYCGT-UHFFFAOYNA-N | |
| 2876391 | |
| C1CCN2CCNCC2C1 |
| C8H16N2 | |
| MFCD01098257 | |
| octahydro-1h-pyrido 1,2-a pyrazine, octahydro-2h-pyrido 1,2-a pyrazine, octahydro-pyrido 1,2-a pyrazine, 1,4-diazabicyclo 4.4.0 decane, octahydro-1h-pyrido 1,2-a piperazine, chembl98511, octahydropyrido 1,2-a pyrazine, 2,3,4,6,7,8,9,9a-octahydro-1h-pyrido 1,2-a pyrazine, +/--1,4-diazabicyclo, octahydropyrido 1,2-a-pyrazine | |
| octahydro-1H-pyrido[1,2-a]pyrazine |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only