missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-1,4-Diazabicyclo[4.4.0]decane, 98+%

Catalog No. p-7034616
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52305MD 250 mg
AAH5230503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52305MD Supplier Thermo Scientific Chemicals Supplier No. H52305MD
Only null left

CAS: 4430-75-5 | C8H16N2 | 140.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4430-75-5
Molecular Formula C8H16N2
Molecular Weight (g/mol) 140.23
MDL Number MFCD01098257
InChI Key ONHPOXROAPYCGT-UHFFFAOYNA-N
Synonym octahydro-1h-pyrido 1,2-a pyrazine, octahydro-2h-pyrido 1,2-a pyrazine, octahydro-pyrido 1,2-a pyrazine, 1,4-diazabicyclo 4.4.0 decane, octahydro-1h-pyrido 1,2-a piperazine, chembl98511, octahydropyrido 1,2-a pyrazine, 2,3,4,6,7,8,9,9a-octahydro-1h-pyrido 1,2-a pyrazine, +/--1,4-diazabicyclo, octahydropyrido 1,2-a-pyrazine
PubChem CID 2876391
IUPAC Name octahydro-1H-pyrido[1,2-a]pyrazine
SMILES C1CCN2CCNCC2C1

Specifications

Boiling Point 68°C to 72°C (3 mmHg)
Quantity 250 mg
UN Number UN3267
Formula Weight 140.23
Percent Purity ≥98%
Chemical Name or Material (+/-)-1,4-Diazabicyclo[4.4.0]decane
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only