missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diazabicyclo[2.2.2]octane bis(sulfur dioxide) adduct, 97%

Catalog No. AC456350010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC456350010 1 g
AC456350050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC456350010 Supplier Thermo Scientific Chemicals Supplier No. 456350010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 119752-83-9 | C6H12N2O4S2 | 240.3 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 119752-83-9
Molecular Formula C6H12N2O4S2
Molecular Weight (g/mol) 240.3
InChI Key RWISEVUOFYXWFO-UHFFFAOYSA-N
Synonym 1,4-diazabicyclo 2.2.2 octane-1,4-diium-1,4-disulfinate, 1,4-diazoniabicyclo 2.2.2 octane-1,4-disulfinate, 1,4-diazabicyclo 2.2.2 octane-1,4-diium-1,4-disulf, 1,4-diazabicyclo 2.2.2 octane bis sulfur dioxide adduct, 1,4-diazoniabicyclo 2.2.2 octane-1,4-diylbis dioxosulfur vi, 1,4-diazabicyclo 2.2.2 octane bis sulfur dioxide adduct sulfur, elemental analysis
PubChem CID 75176251
IUPAC Name 1,4-diazoniabicyclo[2.2.2]octane-1,4-disulfinate
SMILES C1C[N+]2(CC[N+]1(CC2)S(=O)[O-])S(=O)[O-]

Specifications

Melting Point 168.0°C to 172.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min
Packaging Glass Bottle
Quantity 1 g
Formula Weight 240.3
Percent Purity 97%
Chemical Name or Material 1,4-Diazabicyclo[2.2.2]octane bis(sulfur dioxide) adduct
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable solid.
Harmful to
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and k

RUO – Research Use Only