accessibility menu, dialog, popup

UserName

NNSOOSOO

CAS RN 119752-83-9

IUPAC Name: 1,4-diazabicyclo[2.2.2]octane-1,4-diium-1,4-disulfinate
Synonyms: 1,4-diazabicyclo[2.2.2]octane-1,4-diium-1,4-disulfinate
Molecular Weight (g/mol): 240.29
Molecular Formula: C6H12N2O4S2
InChi Key: RWISEVUOFYXWFO-UHFFFAOYSA-N
SMILES: [O-]S(=O)[N+]12CC[N+](CC1)(CC2)S([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)