Learn More
1,4-Diazabicyclo[2.2.2]octane, 97%
CAS: 280-57-9 | C6H12N2 | 112.17 g/mol
Supplier: Thermo Scientific Chemicals 112471000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 1,4-Diazabicyclo[2.2.2]octane | |
| 280-57-9 | |
| 100.0 | |
| White to Yellow | |
| 174.0°C | |
| Authentic | |
| Glass Bottle | |
| MFCD00006689 | |
| 01,1203; 02,99; 04,119; 05,176; 06,157; 07,86; 11,153; 12,155; 13,92; 15,109 | |
| 15,9834 | |
| 20 °C | |
| IMNIMPAHZVJRPE-UHFFFAOYSA-N | |
| 1,4-diazabicyclo[2.2.2]octane | |
| 9237 | |
| 97% |
| 97% | |
| 96.0 | |
| 155.0°C to 160.0°C | |
| 1.1400g/mL | |
| 62°C | |
| 96% min. (GC) | |
| C6H12N2 | |
| 100 g | |
| 1.14 | |
| 1,4-diazabicyclo 2.2.2 octane, triethylenediamine, dabco, dabco 33lv, 1,4-ethylenepiperazine, dabco crystal, teda, texacat td 100, dabco s-25 | |
| Solubility in water: 46g/100mL (26°C). Other solubilities: soluble in most common organic solvents | |
| C1CN2CCN1CC2 | |
| 112.17 | |
| 112.17 | |
| Hygroscopic Crystals |
Chemical Identifiers
| 280-57-9 | |
| 112.17 | |
| IMNIMPAHZVJRPE-UHFFFAOYSA-N | |
| 9237 | |
| C1CN2CCN1CC2 |
| C6H12N2 | |
| MFCD00006689 | |
| 1,4-diazabicyclo 2.2.2 octane, triethylenediamine, dabco, dabco 33lv, 1,4-ethylenepiperazine, dabco crystal, teda, texacat td 100, dabco s-25 | |
| 1,4-diazabicyclo[2.2.2]octane |
Safety and Handling
GHS H Statement
Flammable solid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye damage.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
GHS Signal Word: Danger
EINECSNumber : 205-999-9
RTECSNumber : HM0354200
TSCA : TSCA
RUO – Research Use Only