missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diazabicyclo[2.2.2]octane, 97%

Catalog No. p-101029
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC112470250 25 g Glass Bottle
AC112471000 100 g Glass Bottle
AC112475000 500 g Glass Bottle
AC112470025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC112470250 Supplier Thermo Scientific Chemicals Supplier No. 112470250

CAS: 280-57-9 | C6H12N2 | 112.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 280-57-9
Molecular Formula C6H12N2
Molecular Weight (g/mol) 112.17
MDL Number MFCD00006689
InChI Key IMNIMPAHZVJRPE-UHFFFAOYSA-N
Synonym 1,4-diazabicyclo 2.2.2 octane, triethylenediamine, dabco, dabco 33lv, 1,4-ethylenepiperazine, dabco crystal, teda, texacat td 100, dabco s-25
PubChem CID 9237
IUPAC Name 1,4-diazabicyclo[2.2.2]octane
SMILES C1CN2CCN1CC2

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 155.0°C to 160.0°C
Color White to Yellow
Density 1.1400g/mL
Boiling Point 174.0°C
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 25 g
Fieser 01,1203; 02,99; 04,119; 05,176; 06,157; 07,86; 11,153; 12,155; 13,92; 15,109
Specific Gravity 1.14
Merck Index 15,9834
Solubility Information Solubility in water: 46g/100mL (26°C). Other solubilities: soluble in most common organic solvents
Sublimation Point 20 °C
Formula Weight 112.17
Percent Purity 97%
Physical Form Hygroscopic Crystals
Chemical Name or Material 1,4-Diazabicyclo[2.2.2]octane, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable solid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye damage.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
EINECSNumber 205-999-9
RTECSNumber HM0354200
TSCA TSCA

RUO – Research Use Only