missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diaminobutane Dihydrochloride 98.0+%, TCI America™

Catalog No. p-7126017
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D008125G 25 g
D0081500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D008125G Supplier TCI America Supplier No. D008125G
Only null left

Chemical Identifiers

CAS 333-93-7
Molecular Formula C4H14Cl2N2
Molecular Weight (g/mol) 161.07
MDL Number MFCD00012526
InChI Key XXWCODXIQWIHQN-UHFFFAOYSA-N
Synonym 1,4-diaminobutane dihydrochloride, putrescine dihydrochloride, tetramethylenediamine dihydrochloride, putrescine hydrochloride, 1,4-butanediamine dihydrochloride, unii-x45sur7rhy, 1,4-butanediamine, dihydrochloride, 1,4-butanediamine, hydrochloride, 1,4-diaminobutane, dihydrochloride, butane-1,4-diamine dihydrochloride
PubChem CID 9532
IUPAC Name butane-1,4-diamine;dihydrochloride
SMILES C(CCN)CN.Cl.Cl

Specifications

Color White
Quantity 25 g
Formula Weight 161.07
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Diaminobutane Dihydrochloride
EINECSNumber (2)-0152
RTECSNumber EJ7280000
TSCA Yes