accessibility menu, dialog, popup

UserName

HHClClH2NNH2

CAS RN 333-93-7

IUPAC Name: dihydrogen butane-1,4-diamine dichloride
Synonyms: dihydrogen putrescine dichloride
Molecular Weight (g/mol): 161.07
Molecular Formula: C4H14Cl2N2
InChi Key: XXWCODXIQWIHQN-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].NCCCCN
Molecular Weight (g/mol) 
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (2)
  • (1)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (4)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)