missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diacetoxybenzene 98.0+%, TCI America™

Catalog No. p-7126936
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D180325G 25 g
D1803500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D180325G Supplier TCI America Supplier No. D180325G
Only null left

Chemical Identifiers

CAS 1205-91-0
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00011643
InChI Key AKOGNYJNGMLDOA-UHFFFAOYSA-N
Synonym 1,4-diacetoxybenzene, hydroquinone diacetate, p-diacetoxybenzene, 1,4-benzenediol, diacetate, p-phenylene diacetate, hydroquinone, diacetate, 4-acetyloxy phenyl acetate, 1,4-phenylene diacetate, 1,4-benzenediol, 1,4-diacetate, benzene-1,4-diyl diacetate
PubChem CID 71006
IUPAC Name 4-(acetyloxy)phenyl acetate
SMILES CC(=O)OC1=CC=C(OC(C)=O)C=C1

Specifications

Melting Point 120°C
Color White-Yellow
Quantity 25 g
Formula Weight 194.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Diacetoxybenzene