missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diacetoxybenzene, 98%

Catalog No. p-7025492
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2098714 25 g
AAB2098722 100 g
AAB2098730 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2098714 Supplier Thermo Scientific Chemicals Supplier No. B2098714
Only null left

CAS: 1205-91-0 | C10H10O4 | 194.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1205-91-0
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00011643
InChI Key AKOGNYJNGMLDOA-UHFFFAOYSA-N
Synonym 1,4-diacetoxybenzene, hydroquinone diacetate, p-diacetoxybenzene, 1,4-benzenediol, diacetate, p-phenylene diacetate, hydroquinone, diacetate, 4-acetyloxy phenyl acetate, 1,4-phenylene diacetate, 1,4-benzenediol, 1,4-diacetate, benzene-1,4-diyl diacetate
PubChem CID 71006
IUPAC Name (4-acetyloxyphenyl) acetate
SMILES CC(=O)OC1=CC=C(OC(C)=O)C=C1

Specifications

Melting Point 119°C to 122°C
Quantity 25 g
Beilstein 1912233
Formula Weight 194.19
Percent Purity 98%
Chemical Name or Material 1,4-Diacetoxybenzene
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
EINECSNumber 214-887-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only