missing translation for 'onlineSavingsMsg'
Learn More

1,4-Di-tert-butylbenzene 99.0+%, TCI America™

Catalog No. p-7127112
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
D21115G 5 g
D211125G 25 g
2 options

Catalog No. D21115G

Supplier: TCI America D21115G

Only null left

Chemical Identifiers

CAS 1012-72-2
Molecular Formula C14H22
Molecular Weight (g/mol) 190.33
MDL Number MFCD00008836
InChI Key OOWNNCMFKFBNOF-UHFFFAOYSA-N
Synonym 1,4-di-tert-butylbenzene, p-di-tert-butylbenzene, benzene, p-di-tert-butyl, benzene, 1,4-bis 1,1-dimethylethyl, unii-55ptx4kh71, bis 1,1-dimethylethyl benzene, benzene, bis 1,1-dimethylethyl, 1,4-di-t-butylbenzene, acmc-2097up, 1,4-bis tert-butyl benzene
PubChem CID 13895
IUPAC Name 1,4-ditert-butylbenzene
SMILES CC(C)(C)C1=CC=C(C=C1)C(C)(C)C

Specifications

Melting Point 78°C
Color White
Boiling Point 236°C
Quantity 5 g
Formula Weight 190.33
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Di-tert-butylbenzene
EINECSNumber (3)-0025
RTECSNumber CY8444000
TSCA Yes