missing translation for 'onlineSavingsMsg'
Learn More

1,4-Di-n-butylbenzene, 97+%

Catalog No. AAL1639603
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1639603 1 g
AAL1639609 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1639603 Supplier Thermo Scientific Chemicals Supplier No. L1639603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1571-86-4 | C14H22 | 190.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1571-86-4
Molecular Formula C14H22
Molecular Weight (g/mol) 190.33
MDL Number MFCD01076591
InChI Key VGQOZYOOFXEGDA-UHFFFAOYSA-N
Synonym 1,4-di-n-butylbenzene, benzene, 1,4-dibutyl, p-dibutylbenzol, p-dibutylbenzene, acmc-209dfm, p-di-tert-butyl benzene
PubChem CID 519170
IUPAC Name 1,4-dibutylbenzene
SMILES CCCCC1=CC=C(CCCC)C=C1

Specifications

Melting Point -31°C
Density 0.856
Boiling Point 227°C
Flash Point 110°C (230°F)
Refractive Index 1.489
Quantity 1 g
Beilstein 1858055
Formula Weight 190.33
Percent Purity ≥97%
Chemical Name or Material 1,4-Di-n-butylbenzene
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only