missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Di(4-pyridyl)benzene 95.0+%, TCI America™
$225.56 - $667.33
Chemical Identifiers
| CAS | 113682-56-7 |
|---|---|
| Molecular Formula | C16H12N2 |
| Molecular Weight (g/mol) | 232.29 |
| MDL Number | MFCD00445845 |
| InChI Key | MAWKLXRVKVOYLR-UHFFFAOYSA-N |
| Synonym | 4,4′C-(1,4-Phenylene)bispyridine |
| PubChem CID | 5250442 |
| IUPAC Name | 4-[4-(pyridin-4-yl)phenyl]pyridine |
| SMILES | C1=CC(=CC=N1)C1=CC=C(C=C1)C1=CC=NC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
P1550200MG
|
TCI America
P1550200MG |
200 mg |
Each for $225.56
|
|
|||||
|
P15501G
|
TCI America
P15501G |
1 g |
Each for $667.33
|
|
|||||
Chemical Identifiers
| 113682-56-7 | |
| 232.29 | |
| MAWKLXRVKVOYLR-UHFFFAOYSA-N | |
| 5250442 | |
| C1=CC(=CC=N1)C1=CC=C(C=C1)C1=CC=NC=C1 |
| C16H12N2 | |
| MFCD00445845 | |
| 4,4′C-(1,4-Phenylene)bispyridine | |
| 4-[4-(pyridin-4-yl)phenyl]pyridine |
Specifications
| 113682-56-7 | |
| White-Yellow | |
| MFCD00445845 | |
| 4,4′C-(1,4-Phenylene)bispyridine | |
| C1=CC(=CC=N1)C1=CC=C(C=C1)C1=CC=NC=C1 | |
| 232.29 | |
| 232.29 | |
| Crystalline Powder |
| 193°C | |
| C16H12N2 | |
| 200 mg | |
| MAWKLXRVKVOYLR-UHFFFAOYSA-N | |
| 4-[4-(pyridin-4-yl)phenyl]pyridine | |
| 5250442 | |
| ≥95.0% (GC) | |
| 1,4-Di(4-pyridyl)benzene |
Safety and Handling
TSCA : No