missing translation for 'onlineSavingsMsg'
Learn More

1,4-Di(2-thienyl)-1,4-butanedione 95.0+%, TCI America™

Catalog No. p-7127880
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37121G 1 g
D37125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D37121G Supplier TCI America Supplier No. D37121G

Chemical Identifiers

CAS 13669-05-1
Molecular Formula C12H10O2S2
Molecular Weight (g/mol) 250.33
MDL Number MFCD00208362
InChI Key QJGKCQWQNOPAMG-UHFFFAOYSA-N
PubChem CID 151814
IUPAC Name 1,4-bis(thiophen-2-yl)butane-1,4-dione
SMILES O=C(CCC(=O)C1=CC=CS1)C1=CC=CS1

Specifications

Melting Point 133°C
Quantity 1 g
Formula Weight 250.33
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Di(2-thienyl)-1,4-butanedione