accessibility menu, dialog, popup

UserName

CAS RN 13669-05-1

OOSS

CAS RN 13669-05-1

IUPAC Name: 1,4-bis(thiophen-2-yl)butane-1,4-dione
Synonyms: 1,4-bis(thiophen-2-yl)butane-1,4-dione
Molecular Weight (g/mol): 250.33
Molecular Formula: C12H10O2S2
InChi Key: QJGKCQWQNOPAMG-UHFFFAOYSA-N
SMILES: O=C(CCC(=O)C1=CC=CS1)C1=CC=CS1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results